Learn More
Bochem™ Stainless Steel Beaker
Catalog Number p-7146767
Click to view available options
Capacità (sistema metrico):
100 mL
2000 mL
Bochem™ Stainless Steel Beaker with contact bottom for heating plates and magnetic stirrers.
  • Stainless Steel
  • For use with heating plates and magnetic stirrers
Diametro (sistema metrico) base 50mm
Capacità (sistema metrico) 100 mL
Materiale Stainless Steel (18/10)
Altezza (sistema metrico) 60mm
Include With rim