Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2
Codice prodotto. 15585119 Sfoglia Tutto Fisherbrand Prodotti
Cambia vista
Click to view available options
Peso (sistema metrico):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
200 g
Dimensione della confezione:
1 unità
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Peso (sistema metrico) unitSize
15585119 1 g to 1 kg 1 unità
15565119 200 g 1 unità
15565129 1 g to 5 kg 1 unità
15505109 1 to 100 g 1 unità
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 15585119
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifica
Classificazione E2
Peso (sistema metrico) 1 g to 1 kg
Massa 1 g to 1 kg
Forma Cylindrical