Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F1 with certificate
Sfoglia Tutto Fisherbrand Traceable ProdottiDescrizione
- Class F1 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifica
Specifica
| Classificazione | F1 |
| Peso (sistema metrico) | 1 g to 5 kg |
| Massa | 1 g to 5 kg |