Learn More
Hamilton™ Stainless Steel HC-75 (H+)
Product Code. 10074722
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10074722 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10074722 Supplier Hamilton™ Supplier No. 79476
Spedizione stimata: 04-12-2026
Specifications
Column Capacity 12 meq/column
Particle Size 9 μm
Stationary Phase HC-75 (H+)
Diameter (Metric) Inner 4.1 mm
Diameter (Metric) 4.1 mm