Learn More
De Buyer™ Stainless Steel Hemispherical Bowl
Product Code. 11953684
Change view
Click to view available options
Capacity (Metric):
2.1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11953684 2.1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11953684 Supplier De Buyer™ Supplier No. 3372.20
  • ONLINE EXCLUSIVE PRICE
    55.91€
    32.20€ / pezzi 1
    Risparmia 23.71€
    42% Sconto
Spedizione stimata: 07-12-2026
A professional half-round bowl without base or handles for whisking, heating by bain-marie, storing a mixture and more.
  • Ideal for horizontal mixing
  • Multifunctional, can be used in the fridge, deep freezer, blast chiller and oven
  • Half-round shape helps to incorporate air into a mixture
  • Heavy duty, robust and resistant to deformation
  • Functional and ergonomic open rolled edge, which enables a secure, strong and comfortable grip for dynamic whisking
  • Easy to clean, dishwasher-safe
Specifications
Material Stainless Steel
Capacity (Metric) 2.1 L
Height (Metric) 10.1 cm
Diameter (Metric) 20 cm
Shape Hemispherical, Round Bottom
Product Type Whisking Bowl