Learn More
Alpaco™ Morsetti per tubi in acciaio inox
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Stainless Steel Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
Marca: Alpaco™ INH41
Dettagli aggiuntivi : Peso : 0.12000kg
Descrizione
- Reusable
- Resists pressure
- No lubricant required for assembly
- Smooth clamping to avoid tubing damage
Specifica
36 to 41 mm | |
Acciaio inox | |
10/conf. | |
Fascetta serratubo |