Learn More
Amsta Rostfritt Stainless Steel Lid - Acid Proof For 603/636-2
Product Code. 30285066
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30285066 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30285066 Supplier Amsta Rostfritt Supplier No. 5110132
  • ONLINE EXCLUSIVE PRICE
    29.82€
    25.30€ / pezzi 1
    Risparmia 4.52€
    15% Sconto
Spedizione stimata: 21-12-2026
Highest quality Stainless Steel lid - acid proof For 603/636-2 Amsta bottles and double boiler bottles. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel