Learn More
Amsta Rostfritt Stainless Steel Lid For 603/636-10
Product Code. 30285001
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30285001 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30285001 Supplier Amsta Rostfritt Supplier No. 5010220
  • ONLINE EXCLUSIVE PRICE
    35.15€
    29.60€ / pezzi 1
    Risparmia 5.55€
    15% Sconto
Spedizione stimata: 14-12-2026
Highest Quality Stainless Steel lid For 603/636-10 Amsta bottles and double boiler bottles. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel