Learn More
De Buyer™ Stainless Steel Pastry Bowls
Click to view available options
Volume (Metric):
1 L
3.5 L
4.5 L
5.5 L
7 L
8 L
11.4 L
16.8 L
Height (Metric):
90 mm
100 mm
112 mm
120 mm
125 mm
140 mm
145 mm
150 mm
200 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Pastry Bowl |
| Material | Stainless Steel |
| For Use With (Application) | For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients |
| For Use With (Equipment) | Fridge, deep-freezer, blast-chiller and oven |
| Height (Metric) | 112 mm |
| Quantity | Each |
| Shape | Flat Bottom |
| Sterility | Sterile |
| Volume (Metric) | 3.5 L |