Learn More
Rogo Sampaic™ Stainless Steel Scissors
Product Code. 11706145
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Length (Metric):
110 mm
115 mm
120 mm
140 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
For Use With (Application):
Anatomical
Dissecting
Iris
Metzenbaum
Noyes micro
missing translation for 'unitSize':
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Length (Metric) Tip Style For Use With (Application) missing translation for 'unitSize'
11706145 115 mm Sharp/Sharp Noyes micro Each
11738531 140 mm Round/Round Anatomical Pack of 10
11796135 120 mm Round/Round Dissecting Pack of 10
11768531 110 mm Sharp/Sharp Iris Pack of 10
11786135 140 mm Round/Round Metzenbaum Pack of 10
11748531 140 mm Sharp/Round Anatomical Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
Código de producto. 11706145 missing translation for 'mfr' Rogo Sampaic™ missing translation for 'supplierNo' FMN025
  • ONLINE EXCLUSIVE PRICE
    12.20€
    10.47€ / pezzi 1
    Risparmia 1.73€
    14% Sconto
Ver todas las promociones
In stock
Añadir a la cesta
Edge
Añadir a la cesta
Excellent for a variety of cutting applications in the laboratory. Rogo Sampaic™ Stainless Steel Scissors have a beautiful polished matte finish.
  • Surgical grade stainless steel
  • Available in a variety of styles - Metzenbaum, dissecting, iris, anatomical
Especificaciones
Product Type Micro Scissors
Length (Metric) 115 mm
Material Stainless Steel
For Use With (Application) Noyes micro
Tip Style Sharp/Sharp