Learn More
RETSCH Stainless Steel Test Sieves Intermediate Rings
Product Code. 11964432 Shop All Retsch Products
Change view
Click to view available options
Size:
100 mm dia. x 40 mm H
305 mm dia. x 40 mm H
400 mm dia. x 65 mm H
450 mm dia. x 100 mm H
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11964432 450 mm dia. x 100 mm H Each
10156981 100 mm dia. x 40 mm H Each
10790473 305 mm dia. x 40 mm H Each
10563922 400 mm dia. x 65 mm H Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11964432 Supplier RETSCH Supplier No. 60.935.000450
  • ONLINE EXCLUSIVE PRICE
    228.00€
    207.03€ / pezzi 1
    Risparmia 20.97€
    9% Sconto
Spedizione stimata: 30-03-2027
Specifications
Product Type Test Sieve Intermediate Ring
Material Stainless Steel
Diameter (Metric) 450 mm
Height (Metric) 100 mm
For Use With (Equipment) Vibratory Sieve Shaker AS 450 control
Size 450 mm dia. x 100 mm H