Learn More
Cole-Parmer™ Threaded Reducing Bushings, NPT(M) x NPT(F), Stainless Steel
Cole-Parmer™ Threaded Reducing Bushings, NPT(M) x NPT(F), Stainless Steel offer a more durable, permanent installation.
Marca: Cole-Parmer™ 08539-90
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Sturdy threaded fittings are designed for permanent installation.
- Use with our rigid tubing for your high-pressure applications.
- Tubing sold separately.
Specifica
Rigid tubing for high-pressure applications | |
3/8 in. NPT(M) x 1/8 in. NPT(F) | |
Stainless Steel (316) |
1 Each | |
Threaded Reducing Bushing | |
7500 psi |