Description
Recommended for micro electronics and precision assembly
- Hand finished points for accuracy and excellent tip hardness
- Anti-glare finish reduces eye strain
- Anti-magnetic and acid resistant
Specifications
Specifications
| Lunghezza (sistema metrico) | 14.0 cm |
| Materiale | Stainless Steel |
| N. per confezione | Each |
| Quantità | Ea. |
| Stile | Straight |
| Stile punta | Long Fine |
| Product Type | Tweezers |