Learn More
Bochem™ Stand Base with Adjustable Foot
Product Code. 10578272
Change view
Click to view available options
For Use With (Equipment):
Rods (Thread M10) and Clamps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10578272 Rods (Thread M10) and Clamps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10578272 Supplier Bochem™ Supplier No. 5036
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Spedizione stimata: 10-12-2026
Bochem™ steel varnished stand base has an adjustable foot for stability and is used with M10 rods, rings and clamps for supporting various laboratory apparatus.
  • Steel varnished base DIN 12892
  • Thread M10
  • Adjustable foot
Specifications
Material Varnished Steel
Dimensions (L x W) 250 x 160 mm
Length (Metric) 250 mm
Width (Metric) Base 160 mm
Product Type Stand Base
For Use With (Application) Holding and Supporting Laboratory Apparatus
For Use With (Equipment) Rods (Thread M10) and Clamps