Learn More
Bochem™ Stand Base Rod
Product Code. 10740052
Change view
Click to view available options
Length (Metric):
1500 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10740052 1500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10740052 Supplier Bochem™ Supplier No. 5115
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Spedizione stimata: 11-01-2027
Bochem™ Stand Base Rod are constructed of Stainless Steel and for use with stand bases.
  • Stainless Steel
Specifications
Material Stainless Steel (18/10)
Length (Metric) 1500 mm
Diameter (Metric) Outer 12 mm
Thread Size M10
Description External Threads