Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Adaptor Set, 1/4 in. (3/32 in. wall) to 1/2 in.
Codice prodotto. 17860627 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
1 set
Dimensione della confezione:
pezzi 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17860627 1 set pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17860627 Fornitore Thermo Scientific™ N. del fornitore SH31090.01
  • ONLINE EXCLUSIVE PRICE
    394.00€
    336.18€ / pezzi 10
    Risparmia 57.82€
    14% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 08-03-2027
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
  •  One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Approved for storage at ambient conditions (15°C to 30°C).
Quantità 1 set
Shipping Condition Room Temperature
Stile dei tubi 6.35mm (1/4in) ID x 2.4mm (3/32in) wall to 12.7mm (1/2in) ID x 3.2mm (1/8in) wall C-Flex 374
Tubing Line 1 0.5 m