Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Y Set
Product Code. 17890677 Shop All Thermo Scientific Products
Change view
Click to view available options
Tubing Style:
3 x 12.7mm (1/2 in) ID C-Flex 374
3 x 3.2mm (1/8 in) ID C-Flex 374
3 x 6.35mm (1/4 in) ID C-Flex 374
3 x 9.52mm (3/8 in) ID C-Flex 374
Dimensions:
1/2 in. ID x 1/8 in. wall
1/4 in. ID x 1/16 in. wall
1/4 in. ID x 3/32 in. wall
1/8 in. ID x 1/16 in. wall
3/8 in. ID x 1/8 in. wall
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Style Dimensions Unit Size
17890677 3 x 9.52mm (3/8 in) ID C-Flex 374 3/8 in. ID x 1/8 in. wall Pack of 10
17840627 3 x 3.2mm (1/8 in) ID C-Flex 374 1/8 in. ID x 1/16 in. wall Pack of 10
17850627 3 x 6.35mm (1/4 in) ID C-Flex 374 1/4 in. ID x 1/16 in. wall Pack of 10
17800807 3 x 6.35mm (1/4 in) ID C-Flex 374 1/4 in. ID x 3/32 in. wall Pack of 10
17880627 3 x 12.7mm (1/2 in) ID C-Flex 374 1/2 in. ID x 1/8 in. wall Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Product Code. 17890677 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' SH31095.01
  • ONLINE EXCLUSIVE PRICE
    965.00€
    794.62€ / pezzi 10
    Risparmia 170.38€
    17% Sconto
 Restricted Item
Spedizione stimata: 18-12-2026
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
  •  One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifications
Dimensions 3/8 in. ID x 1/8 in. wall
Content And Storage Approved for storage at ambient conditions (15°C to 30°C).
Tubing Style 3 x 9.52mm (3/8 in) ID C-Flex 374
Tubing Size 2 x 0.51m (20 in) to 0.51m (20 in)