Learn More
Ismatec™ Standard Tubing Connectors, Plastic
Variously sized and shaped tubing connectors. Ismatec™ Standard Tubing Connectors, Plastic are available in different plastic compositions.
Marca:  Ismatec™ ISM513
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10216662
  • 60.28€ / pezzi 10
Spedizione stimata
14-04-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Standard, non-reducing.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Specifica
Specifica
Acetal Resin (POM, Polyoxymethylene)
For 6.0mm I.D. Tubing
6.0mm O.D.
Standard Tubing Connector
10/Pk.
T-shape