Learn More
Pacific Handy Cutter Standard Utility Blade Dispenser
Product Code. 17779027
Change view
Click to view available options
Product Type:
Blade Dispenser
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17779027 Blade Dispenser Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17779027 Supplier Pacific Handy Cutter Supplier No. QBD324
  • ONLINE EXCLUSIVE PRICE
    255.60€
    205.53€ / pezzi 6
    Risparmia 50.07€
    19% Sconto
Spedizione stimata: 16-12-2026
The QBD-324 Blade Dispenser safely dispenses one SB-92 Standard Utility Blade at a time from a durable plastic case by easily pulling the yellow tab.
  • This convenient gravity fed dispenser helps control blade inventory and insures that employees have the proper tools where and when needed to remain productive.
  • The dispenser contains 75 new blades and fits most standard utility knives.
  • Plus, by pairing a PHC Blade Dispenser (SPD-017, QBD-324, or QBD-379) to the front of the PHC Blade Bank you create a blade change station that will increase the safety and efficiency of your staff.
  • By using both items together your employees will be encouraged to properly dispose of loose blades and easily access new replacement blades near their work stations and throughout your facility, ultimately increasing overall productivity.
Specifications
Product Type Blade Dispenser
For Use With (Application) Easily Access New Replacement Blade
For Use With (Equipment) SB-92 Standard Utility Blade
Disposable Single-use
Sterility Non-sterile