missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearic acid, ≥98.5%, MilliporeSigma™ Supelco™
Specifica
Specifications
| CAS | 57-11-4 |
| Pressione del vapore | 1 mmHg (173.7°C) |
| Punto d'infiammabilità | 113°C |
| Shipping Condition | Ambient |
| Forma fisica | Neat |
| Formula lineare | CH3(CH2)16 COOH |
| Numero MDL | MFCD00002752 |
| Punto di fusione | 67°C to 72°C |
| Punti di ebollizione | 361°C |
| Sinonimo | 1-Heptadecanecarboxylic acid; C18:0; Cetylacetic acid; NSC 25956; NSC 261168; Octadecanoic acid; Stearophanic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?