Learn More
Brady™ Maximum Chain Security
Product Code. 15693537 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
8 mm
10 mm
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
15693537 10 mm Pack of 2
15683537 8 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15693537 Supplier Brady™ Supplier No. 805846
  • ONLINE EXCLUSIVE PRICE
    286.00€
    261.50€ / pezzi 2
    Risparmia 24.50€
    8% Sconto
Spedizione stimata: 29-12-2026
More secure than steel cables.
  • Supplied with a fabric sleeve which will protect paintwork and chrome from damage
Specifications
Product Type Chain
Alternate Part Number CHAIN SECURITY 10 MM 2 PK
Diameter (Metric) 10 mm
Quantity 2 Pk.