Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
Sfoglia Tutto Wheaton Science Products ProdottiDescription
- Sterile
- Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
| Sterilità | Sterile |
| N. per custodia | 230 |
| Stile | OmniFlex 3G Straight Plug |
| Product Line | Wheaton CompletePAK |
| Dimensione chiusura | 13 mm |
| Certificazione/Conformità | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |