Learn More
Thermo Scientific™ Sterilin™ Plastic Culture Tubes
Click to view available options
Materiale:
Polypropylene
Polystyrene
Capacità (sistema metrico):
4 mL
12 mL
N. per confezione:
25
N. per custodia:
500
Packungsgröße:
pezzi 500
Beschreibung
Gain versatility in culture applications with these plastic culture tubes. With a patch for labeling and a two-position cap, the tubes are ideal for suspension cell culture.
NOT AVAILABLE IN AMERICA
- Two-position cap—can be left loose for gas exchange or engaged for tight seal
- Large white marker area for easy labeling
- Printed graduations
Ordering Information
- Not available in all geographic regions
Spezifikation
Spezifikation
| Diametro (sistema metrico) | 12 mm |
| Product Type | Culture Tube |
| Tipo di chiusura | Dual-position Cap |
| Materiale | Polystyrene |
| N. per confezione | 25 |
| N. per custodia | 500 |
| Sterilità | Sterile |
| Capacità (sistema metrico) | 4 mL |
| Graduazioni | Printed |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur