Learn More
Sartorius Sterisart NF gamma, Transfersystem
Product Code. 17827960 Shop All Sartorius Products
Change view
Click to view available options
Type:
Tubing system with a dual needle
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17827960 Tubing system with a dual needle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17827960 Supplier Sartorius Supplier No. 16472GBD
  • ONLINE EXCLUSIVE PRICE
    310.00€
    277.55€ / pezzi 10
    Risparmia 32.45€
    10% Sconto
In stock
Add to basket
Edge
Add to basket
The Sterisart™ NF System is a completely disposable technology based on the membrane filtration method, for the sterility testing of pharmaceutical products.
  • Unique Septum Version for aseptic sampling
  • High retention of microbes
  • Low adsorption
  • High mechanical stability
  • Sharp Metal needle for easy penetration
  • Pre-installed color-coded tube clamps
  • Easy-to-read graduated marks
  • User-friendly (several practical adapters available)
  • Product-/lot number identification
  • Huge variety of adapters and more than 20 different unit types
  • Gas-impermeable packaging for protection against sterilants
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Sterility Sterile
For Use With (Application) Sterility Testing
Type Tubing system with a dual needle
For Use With (Equipment) Dual Needle