missing translation for 'onlineSavingsMsg'
Learn More

STF-62247, MedChemExpress

Codice prodotto. 16449706
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
10 mg
25 mg
50 mg
Dimensione della confezione:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16449706 50 mg 50mg
16442646 25 mg 25mg
16413158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16449706 Fornitore Medchem Express N. del fornitore HY100746

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress STF-62247 is an autophagy inducer that selectively cytotoxic to VHL-deficient renal cell carcinoma (IC50 of 0.625 μM and 16 μM in RCC4 and RCC4/VHL cells, respectively).

STF-62247 is an autophagy inducer that selectively cytotoxic to VHL-deficient renal cell carcinoma (IC50 of 0.625 μM and 16 μM in RCC4 and RCC4/VHL cells, respectively).

Specifications

Materiale o nome chimico STF-62247
Forma fisica Solid
CAS 315702-99-9
Colore Yellow
Quantità 50 mg
Note sul grado di purezza Research
Formula molecolare C15H13N3S
Da utilizzare con (applicazione) Cancer-programmed cell death
Informazioni di solubilità DMSO : 50 mg/mL (187.02 mM; Need ultrasonic)
SMILES CC1=CC(NC2=NC(C3=CC=NC=C3)=CS2)=CC=C1
Molecular Weight (g/mol) 267.35
Peso formulazione 267.35
Percent Purity 98.99%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.