Learn More
Lenz Laborglasintrumente Stopcocks, Straight Bore, with Solid Glass Plug
Click to view available options
Foro spina:
2.5 mm
6 mm
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
- Acc. to DIN 12541-1
- With solid glass plug and screwthread retaining device
- One straight bore
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifica
Specifica
| Foro spina | 2.5 mm |
| Materiale | Borosilicate Glass (3.3) |