Description
Chrome-plated steel cork borer set stays sharp.
- Flat blades permit easy boring of lightweight plastic, cork stoppers, and spongy articles.
- Set consists of twelve borers with outside diameters of 4, 6, 7.5, 8.5, 10, 12, 13, 15, 17, 18.5, 19, and 20.5 mm.
Specifications
Specifications
| Diametro (sistema metrico) | 10 mm, 12 mm, 13 mm, 15 mm, 17 mm, 18.5 mm, 19 mm, 20.5 mm, 4 mm, 6 mm, 7.5 mm, 8.5 mm |
| Quantità | 12 Borers/Set |
| Colore | Chrome |
| Materiale | Chrome Plated Steel |
| Product Type | Stopper Hole Boring Set |