missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STPG1 Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 17137608 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
200 μL
Dimensione della confezione:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17137608 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17137608 Fornitore Invitrogen™ N. del fornitore PA5116371

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

STPG1 Polyclonal Antibody for Western Blot

May positively contribute to the induction of apoptosis triggered by O(6)-methylguanine.
TRUSTED_SUSTAINABILITY

Specifica

Antigene STPG1
Applicazioni Western Blot
Classificazione Polyclonal
Coniugato Unconjugated
Formulazione PBS with 0.09% sodium azide; pH 7.4
Gene STPG1
N. accesso geni Q5TH74
Alias gene 4930403G18Rik; 4930555I21Rik; C1orf201; MAPO2; O(6)-methylguanine-induced apoptosis 2; O6-methylguanine-induced apoptosis 2; sperm tail PG rich repeat containing 1; sperm tail PG-rich repeat containing 1; sperm-tail PG-rich repeat containing 1; sperm-tail PG-rich repeat-containing protein 1; STPG1; UPF0490 protein C1orf201; UPF0490 protein C1orf201 homolog
Simboli geni STPG1
Specie ospite Rabbit
Immunogeno KLH conjugated synthetic peptide between 37-70 amino acids from the N-terminal region of human C1orf201.
Metodo di purificazione Antigen affinity chromatography, Protein A
Quantità 200 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 90529
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.