missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-340.5), Invitrogen™
GREENER_CHOICE

Codice prodotto. 11578962
Cambia vista
Click to view available options
Quantità:
500 μg
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11578962 500 μg Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11578962 Fornitore Invitrogen™ N. del fornitore MA110701

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

MA1-10701 detects Streptococci Group A. MA1-10701 has been successfully used in ELISA applications. The MA1-10701 immunogen is UV-inactivated Group A Streptococcus cells.

The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Streptococcus Group A
Applicazioni ELISA
Classificazione Monoclonal
Clone HSA12-340.5
Concentrazione 1.5 mg/mL
Coniugato Unconjugated
Formulazione PBS with no preservative; pH 7.4
Specie ospite Mouse
Immunogeno UV-inactivated Group A Streptococcus cells.
Metodo di purificazione Protein G
Quantità 500 μg
Status giuridico RUO
Primario o secondario Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Forma Liquid
Isotype IgG2a
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.