Learn More
Cytiva Strip Holder
Product Code. 10690216 Shop All Cytiva Products
Change view
Click to view available options
Height (Metric):
7 cm
11 cm
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10690216 11 cm Pack of 6
10301325 7 cm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10690216 Supplier Cytiva Supplier No. 80641630
Spedizione stimata: 07-01-2027
Specifications
Type Strip Holder
Includes Strip Holder Cover, 1 x 60mL Strip Holder Cleaning Solution, Instructions
Quantity 1 Pack
Height (Metric) 11 cm