Learn More
Cole-Parmer™ INC-200 Series Stuart™ Hybridization Incubator/Shaker
Codice prodotto. 11496558
Cambia vista
Click to view available options
Tipo:
Hybridization Oven
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Dimensione della confezione
11496558 Hybridization Oven Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11496558 Fornitore Cole-Parmer™ N. del fornitore 3945405
  • ONLINE EXCLUSIVE PRICE
    4780.00€
    4430.70€ / pezzi 1
    Risparmia 349.30€
    7% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 29-12-2026
INC-200 Series Stuart™ Hybridization Incubator/Shaker is versatile—features both rotisserie and rocking motions.
  • Forced air circulation combined with microprocessor control result in excellent temperature uniformity.
  • Up-and-over door for easy access to the chamber.
  • Use a hybridization incubator when it is vital to maintain the integrity of the sample while heating it to the exact temperature.
  • The incubator is ideal for nucleic acid hybridization applications such as Northern (RNA) blots, Southern (DNA) blots, and Western (protein) blots.
  • This compact incubator comes with two built-in shakers.
  • Rotisserie shaker has a rotational speed from 2 to 10 rpm while the platform shaker has a rocking speed from 5 to 70 oscillations/minute.
  • The gentle rocking action is ideal for the washing stage of most hybridization procedures.
  • Safety features include an overtemperature protection system, plus the power is cut to the motor if the motor overheats or stalls.
  • Stainless steel interior is easy to clean.
  • Double-glazed smoked acrylic and polycarbonate door provides radiation protection.
  • Order a rotisserie that accepts 75 mm OD bottles and hybridization bottles separately.
  • Bottles are made from borosilicate glass and are autoclavable.
Specifica
Capacità (sistema metrico) 20 L
Tipo Hybridization Oven