Learn More
SUCLG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561289
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: FGGIVNCAII ANGITKACRE LELKVPLVVR LEGTNVQEAQ KILNNSGLPI TSAIDLEDAA KKAVASVAKK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
SUCLG2 Polyclonal antibody specifically detects SUCLG2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
SUCLG2 | |
Polyclonal | |
Unconjugated | |
SUCLG2 | |
AF171077; AW556404; D6Wsu120e; GBETA; G-BETA; G-SCS; GTPSCS; GTP-specific succinyl-CoA synthetase beta subunit; GTP-specific succinyl-CoA synthetase subunit beta; hypothetical protein LOC537713; SCS-betaG; Succinate--CoA ligase [GDP-forming] subunit beta, mitochondrial; succinate-CoA ligase GDP-forming beta subunit; succinate-CoA ligase GDP-forming subunit beta; succinate-CoA ligase, GDP-forming, beta subunit; succinate-Coenzyme A ligase, GDP-forming, beta subunit; succinyl-CoA ligase [GDP-forming] subunit beta, mitochondrial; succinyl-CoA ligase, GDP-forming, beta chain, mitochondrial; succinyl-CoA synthetase beta-G chain; succinyl-CoA synthetase, beta-G chain; Suclg2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8801 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96I99 | |
SUCLG2 | |
Recombinant protein corresponding to Human SUCLG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only