Learn More
Sucrose Brix/RI Standards, Paragon Scientific™
Click to view available options
Concentration:
10.00 Brix
12.50 Brix
15.00 Brix
20.00 Brix
25.00 Brix
30.00 Brix
35 Brix
40.00 Brix
45.00 Brix
5.00 Brix
50.00 Brix
55 Brix
7.50 Brix
Unit Size:
15mL
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Primary level refractive index standards traceable to National Standards
- Manufactured in accordance with ICUMSA Methods
- Supplied in tamper evident packs with 6 weeks shelf life
Specifications
Specifications
| Concentration | 10.00 Brix |
| Quantity | 15 mL |
Nominal values quoted