Learn More
Sulfacetamide Sodium Salt Monohydrate, TRC
Specifica
Specifica
| Materiale o nome chimico | Sulfacetamide sodium salt |
| CAS | 6209-17-2 |
| Formula molecolare | C8H9N2O3S.H2O.Na |
| Quantità | 50 g |
| Sinonimo | Sulfacetamide Sodium Monohydrate, N-[(4-Aminophenyl)sulfonyl]acetamide sodium salt hydrate (1:1:1), N-[(4-Aminophenyl)sulfonyl]acetamide monosodium salt monohydrate, N-Sulfanilylacetamide monosodium salt monohydrate, N1-Acetylsulfanilamide sodium monohydrate, Sodium N-sulfanilylacetamide monohydrate, Sodium sulfacetamide monohydrate, Sulamyd sodium monohydrate, Sulfacetamide sodium monohydrate |
| InChI Formula | InChI=1S/C8H10N2O3S.Na.H2O/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;;/h2-5H,9H2,1H3,(H,10,11);;1H2/q;+1;/p-1 |
| SMILES | O.[Na+].CC(=O)[N-]S(=O)(=O)c1ccc(N)cc1 |
| IUPAC Name | sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate |
| Molecular Weight (g/mol) | 254.2387 |
| Peso formulazione | 254.0337 |