Learn More
Sulphate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
BIOPHARMA
Codice prodotto. 12942205 Sfoglia Tutto Fisher Chemical Prodotti
Cambia vista
Click to view available options
Quantità:
100 mL
250 mL
500 mL
Dimensione della confezione:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
12942205 500 mL 500mL
12922205 100 mL 100mL
12932205 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 12942205 Fornitore Fisher Chemical N. del fornitore A 045 500
Please to purchase this item. Need a web account? Register with us today!
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifica
La soluzione contiene H2O
Concentrazione 1 g/L
Quantità 500 mL