missing translation for 'onlineSavingsMsg'
Learn More

SULT2B1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA560411

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15778241

  • 514.00€ / 100µL
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: GLWDTYEDDI SEISQKLPGE YFRYKGVPFP VGLYSLESIS LAENTQDVRD DDIFIITYPK S Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 72%.

SULT2B1 Polyclonal antibody specifically detects SULT2B1 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifica

Specifica

SULT2B1
Polyclonal
Unconjugated
SULT2B1
AI326997; alcohol sulfotransferase; BB173635; HSST2; Hydroxysteroid sulfotransferase 2; RGD:1308882}; ST2B1; sulfotransferase 2B; sulfotransferase 2B1; Sulfotransferase family 2B member 1; sulfotransferase family cytosolic 2B member 1; sulfotransferase family, cytosolic, 2B, member 1; SULT2B; Sult2b1; sult2b1 {ECO:0000312; SULT2B1a
Rabbit
Antigen affinity chromatography
RUO
6820
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O00204
SULT2B1
Recombinant protein corresponding to Human SULT2B1
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only