Learn More
Cellpath Supa Mega Tissue Processing Cassettes
Codice prodotto. p-4908708
Cambia vista
Click to view available options
Colore:
White
Dimensione della confezione:
pezzi 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Colore unitSize
30226233 White pezzi 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30226233 Fornitore Cellpath N. del fornitore EAG010202A
Aggiungi al carrello
Aggiungi al carrello
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
  • Plastic hinged lid
  • Rigid, durable
  • Excellent writing surfaces
  • Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
  • Available in a range of colors
Specifica
Quantità 100/Pk.