Learn More
Cytiva Whatman™ Sieve (Support Screen) for use with MV050 Vacuum Filtration Manifolds
Click to view available options
:
MV050 Sieve (Support Screen)
Descrizione
- Product Type: Sieve (Support Screen)
- Material: Stainless Steel
- For Use with (Equipment): MV050 Vacuum Filtration Manifolds
- For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Monouso | Reusable |
| Da utilizzare con (apparecchiatura) | MV050 Manifold |
| Materiale | Stainless Steel |
| Sterilità | Non-sterile |
| Da utilizzare con (applicazione) | Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries) |
| Modello | MV050/0/03 |
| Linea di prodotti | MV050 |
| Tipo | Sieve (Support Screen) |