Learn More
Smeg™ Supports for Flasks
Designed for use with Smeg™ glassware washing devices. Smeg Washer Supports for Flasks are recommended for flasks with a capacity smaller than 500mL.
Marca: Smeg™ SB25
Dettagli aggiuntivi : Peso : 0.43000kg
Descrizione
- 28-position spring support to hold flasks and beakers
- For use with the standard baskets CS1-1 and CS2
- Capacity of 28 items
- Made of stainless steel
Specifica
Stainless Steel | |
Spring Support | |
28-position for flasks and beakers | |
Standard baskets CS1-1 and CS2 |