Learn More
Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Microtiter Plates, Round V-Bottom, Level 1 Everyday Analysis
Codice prodotto. 15122699
Cambia vista
Click to view available options
Total Volume (Metric) Well:
450 μL
500 μL
N. per confezione:
120 Microplates
120 Plates
20 Plates
Volume (sistema metrico) di lavorazione:
10 to 400 μL
20 to 450 μL
Altezza (sistema metrico):
14.4 mm
14.5 mm
Dimensione della confezione:
pezzi 120
pezzi 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Total Volume (Metric) Well N. per confezione Volume (sistema metrico) di lavorazione Altezza (sistema metrico) Dimensione della confezione
15122699 500 μL 120 Microplates 20 to 450 μL 14.5 mm pezzi 120
15102699 450 μL 20 Plates 10 to 400 μL 14.4 mm pezzi 20
15112699 450 μL 120 Plates 10 to 400 μL 14.4 mm pezzi 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 15122699 Fornitore Thermo Scientific™ N. del fornitore 60180P132
 Articolo soggetto a restrizioni
Spedizione stimata: 07-01-2027
Use SureSTART™ WebSeal™ 96-Well Microtiter Plates, Round V-Bottom, Performance Level 1, for everyday chromatography analysis. They are Specification Certified and conform to ANSI-SLAS 4-2004 standards.
Thermo Scientific™ SureSTART™ Level 1 WebSeal™ 96-Well Round V-Bottom Microtiter Plates are designed for everyday chromatography analysis.
They provide an affordable choice for use with most HPLC/UHPLC instrument types and are ideal for non-regulated laboratories using water and polar solvents.
These 96-well plates are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards. Seal these well plates with SureSTART WebSeal 96-Well Plate Sealing Mats, 8 mm Diameter, Round, Level 1.
Performance Level 1 Plates – for Everyday Analysis
  • Ideal for non-regulated labs that use water and polar solvents
  • Commodity plates, seals, tapes
  • GC tested
  • Suitable for HPLC/UHPLC applications
  • Suitable for biopharmaceutical applications
  • Suitable for QA/QC applications using core detectors
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
  • All Thermo Scientific SureSTART WebSeal well plates are dimensionally verified
Lot Traceable
  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request
About Thermo Scientific WebSeal Well Plates & Mats
Thermo Scientific WebSeal plates and mats are available in three performance levels:
  • Level 1 well plates are for everyday applications and are suitable for non-regulated labs using water and polar solvents
  • Level 2 well plates are for high-throughput applications and are suitable for regulated labs using water and polar solvents
  • Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents
TRUSTED_SUSTAINABILITY
Specifica
Codice a barre No Barcode
Certificato Specification Certified, GC/MS Tested
Matrice 8 x 12
Product Line SureSTART WebSeal Level 1
Colore Clear
Product Type Microplate
Materiale Polypropylene
No. of Wells 96
Volume (sistema metrico) di lavorazione 20 to 450 μL
Sterilità Non-sterile
Trattamento superficiale Non-coated
Total Volume (Metric) Well 500 μL
Forma pozzetto Round, Conical Bottom (V-well)
N. per confezione 120 Microplates
Temperature Range (Metric) -80°C to 121°C
Altezza (sistema metrico) 14.5 mm
Diametro (sistema metrico) pozzetto 8 mm
Plate Size Microtiter