Learn More
Uvex suXXeed GreenCycle Men's Cargo Trousers
GREENER_CHOICE
Catalog Number 17573015
Click to view available options
Size:
42
44
46
48
50
52
54
56
58
60
62
64
90
94
98
102
106
110
The suXXeed GreenCycle planet is the first workwear collection to be fully compostable. All the materials are certified according to the Cradle to Cradle™ principle and are 100% biodegradable.
  • Regular fit
  • Cradle to Cradle certification: gold
  • Recyclable product as part of a circular economy for a clean future
  • Ergonomic cut
  • Elasticated waistband at the back
  • Front fastening with button panel
  • Side pocket on the left with button fastening and inside mobile phone pocket
  • Ruler pocket and pen pocket on the right
  • Back pockets left and right without flap
  • Laser-cut logo on right pocket and on leg
  • Care instructions: wash at 60°C, do not chlorine bleach, dry with reduced thermal stress, iron at medium temperature, can be cleaned with perchlorethylene
  • Machine-washable
Size 102
Sesso Men's
Materiale Cotton, Polyester, Elastane
Colore Anthracite