Learn More
SVL™ Borosilicate Glass Lid for SVL™ Reaction Vessel
Descrizione
- For use with SVL™ reaction vessels
- Borosilicate glass construction available in three styles: - Plain with knob (no necks) to act as a cover - 296-00 - With two necks (one vertical central neck and one vertical side neck) - With five necks as shown in the table below
Specifica
Specifica
| Materiale | Borosilicate Glass |
| Da utilizzare con (apparecchiatura) | SVL™ reaction vessels |
| Altezza (sistema metrico) | 100 mm |
| Product Type | Lid |