Learn More
SVL™ Jacketed Reaction Vessel
Product Code. 15557690
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15557690 1000 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15557690 Supplier SVL Supplier No. 29501
  • ONLINE EXCLUSIVE PRICE
    8746.50€
    7346.42€ / pezzi 5
    Risparmia 1400.08€
    16% Sconto
Spedizione stimata: 05-01-2027
Ideal for temperature controlled reactions.
  • With outer jacket through which a temperature controlling medium can be circulated (maximum temperature of 200°C)
  • Contents of inner chamber can be run off via a Torion PTFE stopcock
  • When used with an SVL™ lid and PTFE covered sealing ring, the assembly is grease free and resistant to chemical attack
Specifications
Capacity (Metric) 1000 mL
Material Glass
For Use With (Application) Ideal for temperature controlled reactions and for circulation of water or oil etc
Product Type Jacketed reaction vessel