Learn More
Micronova Swab Coupon Template Holder with Thick PTFE Template Blade
Codice prodotto. 18840393 Sfoglia Tutto Micronova Prodotti
Cambia vista
Click to view available options
Colore:
White
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Colore Dimensione della confezione
18840393 White Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18840393 Fornitore Micronova N. del fornitore QPSWC16T
  • ONLINE EXCLUSIVE PRICE
    108.00€
    101.37€ / pezzi 1
    Risparmia 6.63€
    6% Sconto
Spedizione stimata: 23-12-2026
Stabilizes sampling templates to define precise, repeatable surface areas
  • Improves operator consistency, supports validation and GMP compliance,differentiates itself by reducing template movement during swab sampling
  • Threaded handle attachment 16 in.
  • Compact HDPE coupon template holder for controlled-area cleaning validation
  • Secure 5 x 5 cm (25 cm2) surface area
  • Threads directly onto STA series universal handles
  • Stable, rigid boundary for repeatable sampling
  • Compatible with standard cleanroom disinfectants and cleaning solutions
  • Lightweight, durable
Specifica
Product Type Swab Holder
Color-coded No
Tipo di maniglia Threaded
Scent Unscented
Sterilità Non-sterile
Autoclavabile Autoclavable
Colore White
Monouso Reusable