Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ stacks.
Secure specimens during processing with Simport Scientific™ Swingsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
Marca:  Simport™ Scientific M518-4T
Dettagli aggiuntivi : Peso : 6.86500kg
 vedi altre versioni di questo prodotto
Codice prodotto. 16595001
  • 313.00€ / pezzi 2000
Spedizione stimata
20-04-2021
Aggiungi al carrello
Descrizione
Descrizione
In Quickload™ stacks
  • 45° Anterior writing area
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifica
Specifica
2000
Green
Acetal