Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ stacks.
Secure specimens during processing with Simport Scientific™ Swingsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
Marca: Simport™ Scientific M518-12T
Dettagli aggiuntivi : Peso : 6.86500kg
Descrizione
In Quickload™ stacks
- 45° Anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifica
2000 | |
Aqua | |
Acetal |