Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ stacks
Secure specimens during processing with Simport Scientific™ Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
Marca: Simport™ Scientific M517-4T
Dettagli aggiuntivi : Peso : 6.38600kg
Descrizione
In Quickload™ stacks
- 45° Anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifica
2000 | |
Green | |
Acetal |