Learn More
Hamilton™ Swivel Union 1/4-28 Threads
Codice prodotto. 11502475
Cambia vista
Click to view available options
Materiale:
CTFE
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Materiale Dimensione della confezione
11502475 CTFE Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11502475 Fornitore Hamilton™ N. del fornitore 35037
  • ONLINE EXCLUSIVE PRICE
    150.00€
    137.98€ / pezzi 1
    Risparmia 12.02€
    8% Sconto
Spedizione stimata: 04-11-2026
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Specification Sheet
  • Connection 1 – Male 1/4-28
  • Connection 2 – Male 1/4-28
  • Fitting Material – CTFE
  • Sales Quantity – 1 each
  • I.D. – 1.5 mm
Specifica
Tipo Male 1/4-28
Materiale CTFE