missing translation for 'onlineSavingsMsg'
Learn More

Syncollin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA561563

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15779371

  • 530.00€ / 100µL
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: TASSLVVAPR CELTVWSRQG KAGKTHKFSA GTYPRLEEYR RGILGDWSNA ISALYCRCP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.

Syncollin Polyclonal antibody specifically detects Syncollin in Human samples. It is validated for Immunohistochemistry.
Specifica

Specifica

Syncollin
Polyclonal
Unconjugated
SYCN
0910001K16Rik; 1810038B08Rik; FLJ27441; INSSA1; insulin synthesis associated 1; insulin synthesis-associated protein 1; Proximal small intestine-specific protein 9; Sip9; Sycn; SYL; syncollin
Rabbit
Antigen affinity chromatography
RUO
342898
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q0VAF6
SYCN
Recombinant protein corresponding to Human Syncollin
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only