Learn More
Corning™ Synthemax™ II Dissolvable Microcarriers
Product Code. 17253084 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17253084 10 g 10g
17233084 1 g 1g
17243084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17253084 Supplier Corning™ Supplier No. 7290
In stock
Corning microcarriers are designed to simplify your scale-up processes.
  • Sterile
  • Ready-to-use
  • Available in closed systems packaging to be used directly with bioreactors
  • Additionally, the microcarriers are surface-treated to enhance cell attachment, maximizing cell yield and viability
  • Consistency – USP Class VI polystyrene material provides a consistent platform
  • Performance – Corning microcarriers are offered untreated or with the following surface treatments
  • Corning Synthemax™ II substrate creates a synthetic surface on the microcarriers for stem cell expansion
  • For use with MSC and iPS/ES applications. (Animal Origin Free)
  • Sterility Assurance Level (SAL) of 10−6
  • Nonpyrogenic
  • Available as closed system solutions
  • Corning microcarriers are available sterile and ready to use in 1 to 10 g vials. This format provides flexibility to select the amount of beads to use.
  • During the mature phase of process development and into manufacturing, closed system packaging would be necessary to maintain sterility and ease bioreactor loading.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) MSC and iPS/ES Applications
Sterility Sterile
Surface Treatment Synthemax™ II
Product Type Microcarrier
Material USP Class VI Polystyrene
Quantity 10 g